Scopoletin (7-Hydroxy-6-methoxycumarin)
Lieferant: EHRENSTORFER GMBH
EHERC16916000EA
184
EUR
EHERC16916000
Scopoletin (7-Hydroxy-6-methoxycumarin)
Scopoletin (7-Hydroxy-6-methoxycumarin)
Formel:
C₁₀H₈O₄ |
MDL:
MFCD00006872 CAS-Nummer: 92-61-5 EINECS: 202-171-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...