N-Acetyl-D-leucin

Lieferant: BACHEM BIOCHEMICA

BACE4001881.0025EA 891 EUR
BACE4001881.0025 BACE4001881.0001 BACE4001881.0005
N-Acetyl-D-leucin
N-Acetyl-D-leucin
Formel: C₈H₁₅NO₃
Molecular Weight: 173,2 g/mol
MDL: MFCD00066069
CAS-Nummer: 19764-30-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR