Bulb pipettes with ground-in piston
Lieferant: Karl Hecht
HECH42555064EA 9.9 EUR
HECH42555064
Bulb pipettes with ground-in piston
Pipetten Vollpipetten
Safety volumetric pipettes with ground-in piston.
  • Interchangeable piston, standard type
  • Fused amber graduation 'ASSISTENT-PERMANENT'
  • Zero at top
  • Calibrated
  • Fire-polished tip