Cyprodinil

Lieferant: EHRENSTORFER GMBH

EHERC11909000EA 77 EUR
EHERC11909000
Cyprodinil
Cyprodinil
Formel: C₁₄H₁₅N₃
CAS-Nummer: 121552-61-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR