Myristicin, Supelco®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

09237-10MG-FEA 57.6 EUR
09237-10MG-F
Myristicin, Supelco®
Myristicin
Formel: C₁₁H₁₂O₃
CAS-Nummer: 607-91-0
EINECS: 210-146-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR