N-Ethylcyclohexylamin, Supelco®

Lieferant: Merck

MET502AEA 41 EUR
MET502A
N-Ethylcyclohexylamin, Supelco®
N-Ethylcyclohexylamin
Formel: C₈H₁₇N
Molecular Weight: 127,23 g/mol
MDL: MFCD00003834
CAS-Nummer: 5459-93-8
EINECS: 226-733-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR