3,4-Dimethylphenyloxamidsäureethylester
Lieferant: Thermo Scientific
MAYBBLT00069.1EA
0
EUR
MAYBBLT00069.1
MAYBBLT00069.250
3,4-Dimethylphenyloxamidsäureethylester
3,4-Dimethylphenyloxamidsäureethylester
Formel:
C₁₂H₁₅NO₃ |
CAS-Nummer:
24451-17-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...