N-Acetyl-D-methionin

Lieferant: BACHEM BIOCHEMICA

BACE4007232.0005EA 99.6 EUR
BACE4007232.0005 BACEF-2300.0025
N-Acetyl-D-methionin
N-Acetyl-D-methionin
Formel: C₇H₁₃NO₃S
Molecular Weight: 191,2 g/mol
MDL: MFCD00069962
CAS-Nummer: 1509-92-8
EINECS: 216-144-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR