6-Methoxy-2-naphthonitril

Lieferant: CHEMPUR

CHMPNRB01645.250EA 0 EUR
CHMPNRB01645.250 MAYBNRB01645.1
6-Methoxy-2-naphthonitril
6-Methoxy-2-naphthonitril
Formel: C₁₂H₉NO
MDL: MFCD00009692
CAS-Nummer: 67886-70-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR