Daminozid (Bernsteinsäuremono(2,2-dimethylhydrazid))

Lieferant: EHRENSTORFER GMBH

EHERC11960000EA 48 EUR
EHERC11960000
Daminozid (Bernsteinsäuremono(2,2-dimethylhydrazid))
Daminozid (Bernsteinsäuremono(2,2-dimethylhydrazid))
Formel: C₆H₁₂N₂O₃
MDL: MFCD00002787
CAS-Nummer: 1596-84-5
EINECS: 216-485-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR