Phenoxyacetonitril, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL499757-5MLEA 12.4 EUR
SIAL499757-5ML
Phenoxyacetonitril, Sigma-Aldrich®
Phenoxyacetonitril
Formel: C₈H₇NO
Molecular Weight: 133,15 g/mol
MDL: MFCD00052017
CAS-Nummer: 3598-14-9
UN: 3276
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR