Phenoxyacetonitril
Lieferant: Thermo Scientific
MAYBRF03205.50EA
0
EUR
MAYBRF03205.50
Phenoxyacetonitril
Phenoxyacetonitril
Formel:
C₈H₇NO Molecular Weight: 133,15 g/mol |
MDL:
MFCD00052017 CAS-Nummer: 3598-14-9 UN: 3276 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...