Leupeptin, hemisulphate
Lieferant: G-Biosciences
786-051EA 354 EUR
786-051
Leupeptin, hemisulphate
Proteine und Peptide
Leupeptin, hemisulphate inhibits serine, plasmin, porcine kallikrein and cysteine proteases, including papain and cathepsin B. Does not inhibit chymotrypsin and thrombin