Inosin

Lieferant: Molekula

MOLEM63240440EA 0 EUR
MOLEM63240440
Inosin
Inosin
Formel: C₁₀H₁₂N₄O₅
Molecular Weight: 268,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00066770
CAS-Nummer: 58-63-9
EINECS: 200-390-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR