Guanosin-5'-diphosphat Dinatriumsalz
Lieferant: Molekula
MOLE78355753-100MGEA
120
EUR
MOLE78355753-100MG
Guanosin-5'-diphosphat Dinatriumsalz
Guanosin-5'-diphosphat Dinatriumsalz
Formel:
C₁₀H₁₃N₅Na₂O₁₁P₂ Molecular Weight: 487,17 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00070121 CAS-Nummer: 7415-69-2 EINECS: 231-026-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...