Chymostatin
Lieferant: G-Biosciences
786-048EA 331 EUR
786-048
Chymostatin
Proteine und Peptide
Chymostatin inhibits serine proteases having a chymotrypsin-like specificity, including α, β γ, and δ chymotrypsin, and most cysteine proteases including cathepsins B, H, L. Soluble in DMSO.