Bacitracin
Lieferant: EHRENSTORFER GMBH
EHERC10418000EA
53
EUR
EHERC10418000
Bacitracin
Bacitracin
Formel:
C₆₆H₁₀₃N₁₇O₁₆S₁ Molecular Weight: 1421,60 g/mol |
MDL:
MFCD00062640 CAS-Nummer: 1405-87-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...