Spermin Tetrahydrochlorid
Lieferant: EHRENSTORFER GMBH
EHERC16972742EA
80
EUR
EHERC16972742
Spermin Tetrahydrochlorid
Spermin Tetrahydrochlorid
Formel:
C₁₀H₃₀Cl₄N₄ Molecular Weight: 348,19 g/mol |
MDL:
MFCD00012914 CAS-Nummer: 306-67-2 EINECS: 206-189-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...