Pentylsilan

Lieferant: SILANE (PART OF CHEMPUR)

SILNSILS12931.1EA 0 EUR
SILNSILS12931.1 SILNSILS12931.25 SILNSILS12931.5
Pentylsilan
Pentylsilane
Formel: C₅H₁₄Si
CAS-Nummer: 10177-98-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR