Immersion thermostats, Tempunit® TU-20
Lieferant: Cole Parmer
Techne®
TECNFTU20DDCEA 1653.33 EUR
TECNFTU20DDC TECNFTU20HDC
Immersion thermostats, Tempunit® TU-20
Umwälzthermostate Offene Umwälzsysteme Thermostatische Regler
"Clip On" Tempunit® thermoregulator designed to be used with Techne® unheated water baths or any other suitable laboratory vessels. They will heat, circulate and safely control the temperature of the liquid in the bath within precise limits. Units have a rugged splashproof case, a powerful pressure pump and external circulation fittings.
  • Four digit setting with a bright LED digital temperature display
  • User adjustable over-temperature cut-out and low liquid level cut-out as standard, TU-20HT has an audible alarm
Maximum pump capacity:
10 l/min, 145 mbar ( TU-20D only)
* Refrigeration or cooling coil required for below ambient cooling. Note the TU-20HT can only be used with the Dip Coolers
Standards: Complies with: DIN 12876
Lieferumfang: Supplied with clamp, TU-20HT supplied with TechneWorks software; other thermoregulators and accessories are available, please contact your local Avantor office for details.