Tubing connectors, SVS, bent
Lieferant: Witeg
WITG0.185.001EA 3.38 EUR
WITG0.185.001
Tubing connectors, SVS, bent
Fittings und Verbinder
Tubing connection consisting of screw cap, hose connection with silicone seal and threaded tubes.
  • Very good chemical resistance
  • Very good temperature resistance