Oxytetracyclin Hydrochlorid

Lieferant: EHRENSTORFER GMBH

EHERC15820000EA 46 EUR
EHERC15820000
Oxytetracyclin Hydrochlorid
Oxytetracyclin Hydrochlorid
Formel: C₂₂H₂₅ClN₂O₉
Molecular Weight: 496,90
MDL: MFCD00135815
CAS-Nummer: 2058-46-0
EINECS: 218-161-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR