Tetracyclin

Lieferant: Molekula

MOLE32100101-25GEA 68.7 EUR
MOLE32100101-25G
Tetracyclin
Tetracyclin
Formel: C₂₂H₂₄N₂O₈
Molecular Weight: 444,43 g/mol
MDL: MFCD00151232
CAS-Nummer: 60-54-8
EINECS: 200-481-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR