Hexazinon
Lieferant: EHRENSTORFER GMBH
EHERC14200000EA
58
EUR
EHERC14200000
Hexazinon
Hexazinon
Formel:
C₁₂H₂₀N₄O₂ |
MDL:
MFCD00055526 CAS-Nummer: 51235-04-2 EINECS: 257-074-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...