Bifenthrin

Lieferant: EHRENSTORFER GMBH

EHERC10584000EA 98 EUR
EHERC10584000
Bifenthrin
Bifenthrin
Formel: C₂₃H₂₂ClF₃O₂
CAS-Nummer: 82657-04-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR