γ-Cyhalothrin
Lieferant: EHRENSTORFER GMBH
EHERC11859500EA
298
EUR
EHERC11859500
γ-Cyhalothrin
γ-Cyhalothrin
Formel:
C₂₃H₁₉ClF₃NO₃ |
CAS-Nummer:
76703-62-3 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...