Procymidon

Lieferant: EHRENSTORFER GMBH

EHERC16310000EA 72 EUR
EHERC16310000
Procymidon
Procymidon
Formel: C₁₃H₁₁Cl₂NO₂
CAS-Nummer: 32809-16-8
EINECS: 251-233-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR