3-Thienylessigsäure

Lieferant: Molekula

MOLEM64100225EA 0 EUR
MOLEM64100225
3-Thienylessigsäure
3-Thienylessigsäure
Formel: C₆H₆O₂S
Molecular Weight: 142,18 g/mol
MDL: MFCD00005473
CAS-Nummer: 6964-21-2
EINECS: 230-166-1
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR