(±)-Dimethenamid

Lieferant: EHRENSTORFER GMBH

EHERC12677500EA 127 EUR
EHERC12677500
(±)-Dimethenamid
(±)-Dimethenamid
Formel: C₁₂H₁₈ClNO₂S
CAS-Nummer: 87674-68-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR