1,5-Isochinolindiol, Millipore®

Lieferant: Merck Millipore Calbiochem

419800-5EA 111 EUR
419800-5
1,5-Isochinolindiol, Millipore®
1,5-Isochinolindiol
Formel: C₉H₇NO₂
Molecular Weight: 161,16 g/mol
MDL: MFCD00006900
CAS-Nummer: 5154-02-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR