Cartap Hydrochlorid

Lieferant: EHRENSTORFER GMBH

EHERC11050000EA 106 EUR
EHERC11050000
Cartap Hydrochlorid
Cartap Hydrochlorid
Formel: C₇H₁₆ClN₃O₂S₂
CAS-Nummer: 15263-53-3
EINECS: 239-309-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR