Amitraz
Lieferant: EHRENSTORFER GMBH
EHERC10230000EA
77
EUR
EHERC10230000
Amitraz
Amitraz
Formel:
C₁₉H₂₃N₃ |
MDL:
MFCD00069396 CAS-Nummer: 33089-61-1 EINECS: 251-375-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...