Ferbam (Eisentris(dimethyldithiocarbamat))

Lieferant: EHRENSTORFER GMBH

EHERC13640000EA 72 EUR
EHERC13640000
Ferbam (Eisentris(dimethyldithiocarbamat))
Ferbam (Eisentris(dimethyldithiocarbamat))
Formel: C₉H₁₈FeN₃S₆
MDL: MFCD00067269
CAS-Nummer: 14484-64-1
EINECS: 238-484-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR