Ametryn

Lieferant: EHRENSTORFER GMBH

EHERC10150000EA 66 EUR
EHERC10150000
Ametryn
Ametryn
Formel: C₉H₁₇N₅S
CAS-Nummer: 834-12-8
EINECS: 212-634-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR