Chlorthal-dimethyl

Lieferant: EHRENSTORFER GMBH

EHERC11620000EA 66 EUR
EHERC11620000
Chlorthal-dimethyl
Chlorthal-dimethyl
Formel: C₁₀H₆Cl₄O₄
Molecular Weight: 331,97
MDL: MFCD00014902
CAS-Nummer: 1861-32-1
EINECS: 217-464-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR