Eswabs collection kit
Lieferant: COPAN Diagnostics
COPA481CEEA 0 EUR
COPA481CE COPA480CE
Eswabs collection kit
Tupfer Tupfer für die Probenahme
Universal open platform for bacterial culture and the preservation of DNA, RNA and antigens of diverse microorganisms. Features unique molded breakpoint with coloured indication mark and swab capture cap.
  • Sample rapidly elutes into liquid phase
  • Provides 1 ml for multitest analysis
  • Maintains viability of aerobic, anaerobic and fastidious bacteria
Preserves nucleic acids and antigens of bacteria, viruses and Chlamydia.
Standards: Certified DNase and RNase free.
Lieferumfang: 50 kits per shelf pack – 500 kits per box.
Bitte beachten *Produkte auf dieser Seite sind außerhalb des Anwendungsbereichs der EU Richtlinie 2019/904