Urine collection swabs, UriSwab®
Lieferant: COPAN Diagnostics
COPA800CEA 740 EUR
COPA800C
Urine collection swabs, UriSwab®
Swabs for the sampling of urine specimens from the collection site to the testing laboratory.
  • Use directly onto the sponge or dip in the sample
  • Chemically bonded preservatives
  • Activation on contact with urine
  • Specimen can be manually extracted by shaking the device
Advanced system specifically designed for the collection, transport and preservation of urine.
According to the testing executed as indicated in the second edition of the "Quality Control of Microbiological Transport Systems" standard, in the laboratory, UriSwab® can be used to process urine specimens for the cultivation of uropathogenic bacteria and yeasts.
Sample and device storage temperature: 2 to 25 °C
Standards: Compliance with CLSI M40-A2 standard: Ch. 9.2 Urine Culture Containers With Preservative.