Sicherheitsschuhe, zum Schnüren,, Origin
Lieferant: Uvex
UVEX8450.9/45EA 103 EUR
UVEX8450.9/45 UVEX8450.9/43 UVEX8450.9/46 UVEX8450.9/44 UVEX8450.9/42 UVEX8450.9/40 UVEX8450.9/41 UVEX8450.9/47
Sicherheitsschuhe, zum Schnüren,, Origin
Extra wide robust safety shoe made from water-repellent nappa leather, padded tongue and collar and a PUR/rubber sole resistant to chemicals, cuts and temperatures from –20 to +300 °C.
  • Breathable textile lining
  • uvex 3D hydroflex® foam insole for exceptional shock absorption in the heel and under the front of the foot
  • Anti-slip sole with shock absorption at heel and front of the foot
  • Protective steel toecap (200 J)
  • Puncture resistant steel midsole
Colour: Black
Width: 11
Standards: EN ISO 20345:2011; S2, CI, HI, HRO, SRC