Captafol (1,2,3,6-Tetrahydro-N-(1,1,2,2-tetrachlorethylthio)phthalimid)

Lieferant: EHRENSTORFER GMBH

EHERC10950000EA 63 EUR
EHERC10950000
Captafol (1,2,3,6-Tetrahydro-N-(1,1,2,2-tetrachlorethylthio)phthalimid)
Captafol (1,2,3,6-Tetrahydro-N-(1,1,2,2-tetrachlorethylthio)phthalimid)
Formel: C₁₀H₉Cl₄NO₂S
Molecular Weight: 349,06 g/mol
CAS-Nummer: 2425-06-1
EINECS: 219-363-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR