Chlorbenzilat (4,4'-Dichlorbenzilsäureethylester)

Lieferant: EHRENSTORFER GMBH

EHERC11390000EA 62 EUR
EHERC11390000
Chlorbenzilat (4,4'-Dichlorbenzilsäureethylester)
Chlorbenzilat (4,4'-Dichlorbenzilsäureethylester)
Formel: C₁₆H₁₄Cl₂O₃
Molecular Weight: 325,19 g/mol
CAS-Nummer: 510-15-6
EINECS: 208-110-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR