2,4,5-T-Methylester (2,4,5-Trichlorphenoxyessigsäuremethylester)
Lieferant: EHRENSTORFER GMBH
EHERC17125000EA
46
EUR
EHERC17125000
2,4,5-T-Methylester (2,4,5-Trichlorphenoxyessigsäuremethylester)
2,4,5-T-Methylester (2,4,5-Trichlorphenoxyessigsäuremethylester)
Formel:
C₉H₇Cl₃O₃ Molecular Weight: 269,51 g/mol |
CAS-Nummer:
1928-37-6 EINECS: 217-669-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...