Chlorpropham (Isopropyl 3-chlorocarbanilat)

Lieferant: EHRENSTORFER GMBH

EHERC11580000EA 49 EUR
EHERC11580000
Chlorpropham (Isopropyl 3-chlorocarbanilat)
Chlorpropham (Isopropyl 3-chlorocarbanilat)
Formel: C₁₀H₁₂ClNO₂
Molecular Weight: 213,66 g/mol
MDL: MFCD00037108
CAS-Nummer: 101-21-3
EINECS: 202-925-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR