Flurenol-Butylester (9-Hydroxy-9H-fluoren-9-carbonsäurebutylester)

Lieferant: EHRENSTORFER GMBH

EHERC13812000EA 94 EUR
EHERC13812000
Flurenol-Butylester (9-Hydroxy-9H-fluoren-9-carbonsäurebutylester)
Flurenol-Butylester (9-Hydroxy-9H-fluoren-9-carbonsäurebutylester)
Formel: C₁₈H₁₈O₃
CAS-Nummer: 2314-09-2
EINECS: 219-011-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR