Trichlorfon (Dimethyl-2,2,2-trichlor-1-hydroxyethylphosphonat)

Lieferant: EHRENSTORFER GMBH

EHERC17680000EA 54 EUR
EHERC17680000
Trichlorfon (Dimethyl-2,2,2-trichlor-1-hydroxyethylphosphonat)
Trichlorfon (Dimethyl-2,2,2-trichlor-1-hydroxyethylphosphonat)
Formel: C₄H₈Cl₃O₄P
Molecular Weight: 257,44 g/mol
CAS-Nummer: 52-68-6
EINECS: 200-149-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR