2,4,5-T (2,4,5-Trichlorphenoxyessigsäure)

Lieferant: EHRENSTORFER GMBH

EHERC17100000EA 54 EUR
EHERC17100000
2,4,5-T (2,4,5-Trichlorphenoxyessigsäure)
2,4,5-T (2,4,5-Trichlorphenoxyessigsäure)
Formel: C₈H₅Cl₃O₃
Molecular Weight: 255,48 g/mol
MDL: MFCD00004301
CAS-Nummer: 93-76-5
EINECS: 202-273-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR