2-Amino-1,3,4-thiadiazol, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ quality level = MQ100

SIAL258881-5GEA 55.9 EUR
SIAL258881-5G
2-Amino-1,3,4-thiadiazol, Sigma-Aldrich®
2-Amino-1,3,4-thiadiazole
Formel: C₂H₃N₃S
Molecular Weight: 101,13 g/mol
MDL: MFCD00003107
CAS-Nummer: 4005-51-0
EINECS: 223-657-7
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR