1-Amino-3,3-diethoxypropan

Lieferant: ABCR GMBH

ABCRAB131351.0025EA 0 EUR
ABCRAB131351.0025
1-Amino-3,3-diethoxypropan
1-Amino-3,3-diethoxypropane
Formel: C₇H₁₇NO₂
MDL: MFCD00142581
CAS-Nummer: 41365-75-7
UN: 2735
ADR: 8,I

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR