Dextran, M.W. ~ 40,000

EDQMD0735000EA 235 EUR
EDQMD0735000
Dextran, M.W. ~ 40,000
Dextran
Formel: (C₆H₁₀O₅)n
MDL: MFCD00130935
CAS-Nummer: 9004-54-0
EINECS: 232-677-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR