Tribromethylen

Lieferant: ABCR GMBH

ABCRAB135145.0025EA 0 EUR
ABCRAB135145.0025
Tribromethylen
Tribromethylen
Formel: C₂HBr₃
Molecular Weight: 264,74 g/mol
MDL: MFCD00039164
CAS-Nummer: 598-16-3
EINECS: 209-920-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR