Filter tips
Lieferant: Greiner bio-one
732-0133EA 0 EUR
732-0133 732-0129
Filter tips
Pipettenspitzen
Recommended for work with DNA and RNA, and when handling radioactive material.
  • High quality PP with PE filter, sterile, graduated, autoclavable
  • Prevent contamination with liquids and aerosols during the pipetting process
  • Free of detectable DNase, RNase and human DNA, non pyrogenic, non cytotoxic